Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Benzhydrylazetidin-3-yl)methanol 98%

Supplier: AMBEED, INC
Warning

A296726-10G A296726-1G A296726-5G
76787-646EA 21 USD InStock
76787-646 76787-648 76787-650
(1-Benzhydrylazetidin-3-yl)methanol 98%
(1-Benzhydrylazetidin-3-yl)methanol
Precaución: for Research Use Only


Formula: C₁₇H₁₉NO
MDL Number: MFCD03407522
CAS Number: 72351-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty