Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1-Tris(hydroxymethyl)ethane 95%

Supplier: AMBEED, INC
Warning

A297794-1000G A297794-100G A297794-25G A297794-500G
76787-660EA 23 USD InStock
76787-660 76787-662 76787-664 76787-666
1,1,1-Tris(hydroxymethyl)ethane 95%
1,1,1-Tris(hydroxymethyl)ethane
Precaución: for Research Use Only


Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 287
MDL Number: MFCD00004687
CAS Number: 77-85-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty