Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2,3-difluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A299329-100G A299329-1G A299329-25G A299329-5G
76787-690EA 153 USD InStock
76787-690 76787-692 76787-694 76787-696
Ethyl-2,3-difluorobenzoate 97%
Ethyl-2,3-difluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₈F₂O₂
MDL Number: MFCD06204337
CAS Number: 773134-65-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty