Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-1-Octenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A301542-1G A301542-250MG A301542-5G
76787-750EA 218 USD InStock
76787-750 76787-752 76787-754
trans-1-Octenylboronic acid pinacol ester 98%
trans-1-Octenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₇BO₂
MW: 238.18 g/mol
MDL Number: MFCD03453667
CAS Number: 83947-55-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty