Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-1-methyl-1H-benzimidazole 95%

Supplier: AMBEED, INC
Warning

A311530-10G A311530-1G A311530-250MG A311530-5G
76787-926EA 143 USD InStock
76787-926 76787-928 76787-930 76787-932
6-Bromo-1-methyl-1H-benzimidazole 95%
6-Bromo-1-methyl-1H-benzimidazole
Precaución: for Research Use Only


Formula: C₈H₇BrN₂
MDL Number: MFCD09027271
CAS Number: 53484-16-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty