Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-fluoro-5-methoxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A312782-100MG A312782-1G A312782-250MG
76787-956EA 11 USD InStock
76787-956 76787-958 76787-960
4-Amino-2-fluoro-5-methoxybenzoic acid 95%
4-Amino-2-fluoro-5-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₈FNO₃
MW: 185.15 g/mol
MDL Number: MFCD11849929
CAS Number: 1001346-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty