Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2R)-cis-2-Aminocyclohexanol hydrochloride 98%

Supplier: AMBEED, INC
Warning

A314196-10G A314196-250MG A314196-25G A314196-5G
76787-974EA 286 USD InStock
76787-974 76787-976 76787-978 76787-980
(1S,2R)-cis-2-Aminocyclohexanol hydrochloride 98%
(1S,2R)-cis-2-Aminocyclohexanol hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClNO
MW: 151.6345 g/mol
MDL Number: MFCD11618003
CAS Number: 200352-28-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty