Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sudan red 7B (fat red bluish, solvent red 19, fat red 7B) 98%

Supplier: AMBEED, INC
Warning

A320489-100G A320489-1G A320489-25G A320489-5G
76788-050EA 569 USD InStock
76788-050 76788-052 76788-054 76788-056
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B) 98%
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Precaución: for Research Use Only


Formula: C₂₄H₂₁N₅
MDL Number: MFCD00003904
CAS Number: 6368-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty