Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-chloro-1H-indazole 98%

Supplier: AMBEED, INC
Danger

A324676-1G A324676-250MG A324676-5G
76788-100EA 38 USD InStock
76788-100 76788-102 76788-104
4-Bromo-3-chloro-1H-indazole 98%
4-Bromo-3-chloro-1H-indazole
Precaución: for Research Use Only


Formula: C₇H₄BrClN₂
MDL Number: MFCD09026985
CAS Number: 1000343-46-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty