Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,6-Dichloro-3-nitrophenyl)ethanone 95%

Supplier: AMBEED, INC
Danger

A326351-100MG A326351-1G A326351-250MG
76788-132EA 35 USD InStock
76788-132 76788-134 76788-136
1-(2,6-Dichloro-3-nitrophenyl)ethanone 95%
1-(2,6-Dichloro-3-nitrophenyl)ethanone
Precaución: for Research Use Only


Formula: C₈H₅Cl₂NO₃
MW: 234.03 g/mol
MDL Number: MFCD17170414
CAS Number: 223785-76-0
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty