Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid 97%

Supplier: AMBEED, INC
Danger

A333395-100MG A333395-250MG A333395-1G A333395-5G
76788-198EA 14 USD InStock
76788-198 76788-200 77668-510 77668-512
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid 97%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₅ClN₂O₂
MDL Number: MFCD09880128
CAS Number: 1000340-37-3
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty