Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodo-3-nitrobenzamide 98%

Supplier: AMBEED, INC
Warning

A352626-100MG A352626-10MG A352626-1G A352626-250MG A352626-25G A352626-25MG A352626-50MG A352626-5G A352626-1MG A352626-5MG
76788-416EA 15 USD InStock
76788-416 76788-418 76788-420 76788-422 76788-424 76788-426 76788-428 76788-430 77668-624 77668-626
4-Iodo-3-nitrobenzamide 98%
4-Iodo-3-nitrobenzamide
Precaución: for Research Use Only


Formula: C₇H₅IN₂O₃
MW: 292.0307 g/mol
MDL Number: MFCD11110639
CAS Number: 160003-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty