Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-(chloromethyl)-4-fluorobenzene 97%

Supplier: AMBEED, INC
Danger

A362960-100MG A362960-1G A362960-250MG A362960-5G
76788-580EA 28 USD InStock
76788-580 76788-582 76788-584 77668-682
1-Bromo-2-(chloromethyl)-4-fluorobenzene 97%
1-Bromo-2-(chloromethyl)-4-fluorobenzene
Precaución: for Research Use Only


Formula: C₇H₅BrClF
MW: 223.48 g/mol
MDL Number: MFCD09263441
CAS Number: 857276-61-0
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty