Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-2H-1,2,3-triazole 95%

Supplier: AMBEED, INC
Warning

A364983-250MG A364983-25G A364983-5G A364983-1G
76788-602EA 6 USD InStock
76788-602 76788-604 76788-606 77594-084
4-Nitro-2H-1,2,3-triazole 95%
4-Nitro-2H-1,2,3-triazole
Precaución: for Research Use Only


Formula: C₂H₂N₄O₂
MW: 114.06 g/mol
MDL Number: MFCD03035233
CAS Number: 84406-63-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty