Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetyl-L-prolinamide 98%

Supplier: AMBEED, INC
Warning

A372547-1G A372547-250MG A372547-25G A372547-5G
76788-688EA 19 USD InStock
76788-688 76788-690 76788-692 76788-694
N-Acetyl-L-prolinamide 98%
N-Acetyl-L-prolinamide
Precaución: for Research Use Only


Formula: C₇H₁₂N₂O₂
MDL Number: MFCD00037340
CAS Number: 16395-58-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty