Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-hydroxyphenylacetic acid 97%

Supplier: AMBEED, INC
Warning

A376984-10G A376984-1G A376984-250MG A376984-25G A376984-5G
76788-752EA 51 USD InStock
76788-752 76788-754 76788-756 76788-758 76788-760
5-Bromo-2-hydroxyphenylacetic acid 97%
5-Bromo-2-hydroxyphenylacetic acid
Precaución: for Research Use Only


Formula: C₈H₇BrO₃
MDL Number: MFCD16620636
CAS Number: 38692-72-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty