Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfadoxin 98%

Supplier: AMBEED, INC
Warning

A377369-50MG A377369-5G A377369-10MG
76788-762EA 6 USD InStock
76788-762 76788-764 77668-746
Sulfadoxin 98%
Sulfadoxin
Precaución: for Research Use Only


Formula: C₁₂H₁₄N₄O₄S
MW: 310.33 g/mol
MDL Number: MFCD00792890
CAS Number: 2447-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty