Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazolo[3,4-b]pyrazin-3-amine 95%

Supplier: AMBEED, INC
Warning

A396823-100MG A396823-1G A396823-250MG A396823-5G
76788-904EA 10 USD InStock
76788-904 76788-906 76788-908 76788-910
1H-Pyrazolo[3,4-b]pyrazin-3-amine 95%
1H-Pyrazolo[3,4-b]pyrazin-3-amine
Precaución: for Research Use Only


Formula: C₅H₅N₅
MDL Number: MFCD09909663
CAS Number: 81411-64-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty