Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloroquinoline-7-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A399880-100MG A399880-1G A399880-250MG A399880-5G
76788-914EA 22 USD InStock
76788-914 76788-916 76788-918 76788-920
4-Chloroquinoline-7-carboxylic acid 98%
4-Chloroquinoline-7-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₆ClNO₂
MDL Number: MFCD11518966
CAS Number: 49713-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty