Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl 1,2,3,6-tetrahydropyridine-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A405296-10G A405296-1G A405296-25G A405296-5G
76788-984EA 31 USD InStock
76788-984 76788-986 76788-988 77601-786
Benzyl 1,2,3,6-tetrahydropyridine-1-carboxylate 98%
Benzyl 1,2,3,6-tetrahydropyridine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₅NO₂
MDL Number: MFCD07366998
CAS Number: 66207-23-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty