Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Fluoro-4-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A407564-100MG A407564-1G A407564-250MG
76788-990EA 25 USD InStock
76788-990 76788-992 76788-994
(2-Fluoro-4-(trifluoromethyl)phenyl)acetic acid 98%
(2-Fluoro-4-(trifluoromethyl)phenyl)acetic acid
Precaución: for Research Use Only


Formula: C₉H₆F₄O₂
MDL Number: MFCD00061186
CAS Number: 209991-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty