Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-((Benzyloxy)carbonyl)piperidine-2-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A420537-100MG A420537-1G A420537-250MG A420537-5G
76789-136EA 7 USD InStock
76789-136 76789-138 76789-140 76789-142
(R)-1-((Benzyloxy)carbonyl)piperidine-2-carboxylic acid 98%
(R)-1-((Benzyloxy)carbonyl)piperidine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₄
MDL Number: MFCD01863738
CAS Number: 28697-09-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty