Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydrofurfuryl propionate 97%

Supplier: AMBEED, INC
Warning

A423979-100G A423979-25G A423979-500G A423979-5G
76789-160EA 66 USD InStock
76789-160 76789-162 76789-164 76789-166
Tetrahydrofurfuryl propionate 97%
Tetrahydrofurfuryl propionate
Precaución: for Research Use Only


Formula: C₈H₁₄O₃
MDL Number: MFCD00047080
CAS Number: 637-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty