Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-bromopyridine 97%

Supplier: AMBEED, INC
Warning

A428862-1G A428862-250MG A428862-25G A428862-5G
76789-204EA 6 USD InStock
76789-204 76789-206 76789-208 76789-210
2-Benzyloxy-5-bromopyridine 97%
2-Benzyloxy-5-bromopyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₀BrNO
MW: 264.12 g/mol
MDL Number: MFCD09037462
CAS Number: 83664-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty