Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Valethamate bromide 98%

Supplier: AMBEED, INC
Warning

A439776-1G A439776-250MG A439776-5G
76789-258EA 87 USD InStock
76789-258 76789-260 76789-262
Valethamate bromide 98%
Valethamate bromide
Precaución: for Research Use Only


Formula: C₁₉H₃₂BrNO₂
MDL Number: MFCD00031617
CAS Number: 90-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty