Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(2-aminophenoxy)piperidine-1-carboxylate 95+%

Supplier: AMBEED, INC
Warning

A442857-100MG A442857-1G
76789-278EA 39 USD InStock
76789-278 76789-280
tert-Butyl 4-(2-aminophenoxy)piperidine-1-carboxylate 95+%
tert-Butyl 4-(2-aminophenoxy)piperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₆H₂₄N₂O₃
MW: 292.37 g/mol
MDL Number: MFCD05865139
CAS Number: 690632-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty