Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsulfoxonium chloride 97%

Supplier: AMBEED, INC
Warning

A444377-100G A444377-25G A444377-500G A444377-5G
76789-294EA 49 USD InStock
76789-294 76789-296 76789-298 76789-300 76789-302
Trimethylsulfoxonium chloride 97%
Trimethylsulphoxonium chloride
Precaución: for Research Use Only


Formula: C₃H₉ClOS
MDL Number: MFCD00011898
CAS Number: 5034-06-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty