Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydrofuran-3-carbaldehyde 98%

Supplier: AMBEED, INC
Danger

A448291-1G A448291-5G A448291-25G
76789-334EA 9 USD InStock
76789-334 76789-336 77551-350
Tetrahydrofuran-3-carbaldehyde 98%
Tetrahydrofuran-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD02101976
CAS Number: 79710-86-4
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty