Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triphenylmethane 98%

Supplier: AMBEED, INC
Warning

A457629-100G A457629-25G A457629-500G A457629-1000G
76789-436EA 45 USD InStock
76789-436 76789-438 76789-440 76789-442 77669-056
Triphenylmethane 98%
Triphenylmethane
Precaución: for Research Use Only


Formula: C₁₉H₁₆
MW: 244.34 g/mol
Boiling Pt: 359
MDL Number: MFCD00004763
CAS Number: 519-73-3
Merck Index: 12,09871
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty