Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans,trans-1,4-Diphenyl-1,3-butadiene 98%

Supplier: AMBEED, INC
Warning

A461507-100G A461507-10G A461507-1G A461507-25G A461507-5G
76789-484EA 147 USD InStock
76789-484 76789-486 76789-488 76789-490 76789-492
trans,trans-1,4-Diphenyl-1,3-butadiene 98%
trans,trans-1,4-Diphenyl-1,3-butadiene
Precaución: for Research Use Only


Formula: C₁₆H₁₄
MDL Number: MFCD00004791
CAS Number: 538-81-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty