Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isoquinolineboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A465529-1G A465529-250MG A465529-25G A465529-5G
76789-512EA 42 USD InStock
76789-512 76789-514 76789-516 76789-518
4-Isoquinolineboronic acid pinacol ester 96%
4-Isoquinolineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₁₈BNO₂
MW: 255.1199 g/mol
MDL Number: MFCD05663891
CAS Number: 685103-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty