Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Bromo-β-nitrostyrene 95%

Supplier: AMBEED, INC
Warning

A473439-1G A473439-250MG A473439-10G
76789-590EA 29 USD InStock
76789-590 76789-592 77669-124
trans-4-Bromo-β-nitrostyrene 95%
trans-4-Bromo-β-nitrostyrene
Precaución: for Research Use Only


Formula: C₈H₆BrNO₂
MDL Number: MFCD00191856
CAS Number: 5153-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty