Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)benzhydrol 97%

Supplier: AMBEED, INC
Warning

A477386-1G A477386-25G A477386-5G
76789-636EA 25 USD InStock
76789-636 76789-638 76789-640
4-(Trifluoromethyl)benzhydrol 97%
4-(Trifluoromethyl)benzhydrol
Precaución: for Research Use Only


Formula: C₁₄H₁₁F₃O
MDL Number: MFCD00014399
CAS Number: 395-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty