Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methylbenzyl bromide 98%

Supplier: AMBEED, INC
Danger

A481772-1G A481772-250MG A481772-25G A481772-5G
76789-692EA 32 USD InStock
76789-692 76789-694 76789-696 76789-698
4-Fluoro-2-methylbenzyl bromide 98%
4-Fluoro-2-methylbenzyl bromide
Precaución: for Research Use Only


Formula: C₈H₈BrF
MDL Number: MFCD03701059
CAS Number: 862539-91-1
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty