Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triethylene glycol monomethyl ether 95%

Supplier: AMBEED, INC
Warning

A487389-100G A487389-25G A487389-500G A487389-1000G
76789-746EA 8 USD InStock
76789-746 76789-748 76789-750 77669-178
Triethylene glycol monomethyl ether 95%
Triethylene glycol monomethyl ether
Precaución: for Research Use Only


Formula: C₇H₁₆O₄
MW: 164.2 g/mol
MDL Number: MFCD00002875
CAS Number: 112-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty