Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc undecylenate 98%

Supplier: AMBEED, INC
Danger

A491938-100MG A491938-1G A491938-5G
76789-812EA 5 USD InStock
76789-812 76789-814 76789-816
Zinc undecylenate 98%
Zinc undecylenate
Precaución: for Research Use Only


Formula: C₂₂H₃₈O₄Zn
MDL Number: MFCD00067246
CAS Number: 557-08-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty