Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium sucrose octasulfate 98%

Supplier: AMBEED, INC
Warning

A493352-100MG A493352-1G A493352-250MG A493352-5G
76789-824EA 33 USD InStock
76789-824 76789-826 76789-828 76789-830
Sodium sucrose octasulfate 98%
Sodium sucrose octasulfate
Precaución: for Research Use Only


Formula: C₁₂H₁₄Na₈O₃₅S₈
MDL Number: MFCD00933758
CAS Number: 74135-10-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty