Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1,5,5,5-Hexamethyl-3-phenyl-3-(trimethylsilyloxy)trisiloxane 98%

Supplier: AMBEED, INC
Warning

A496842-100G A496842-25G
76789-886EA 66 USD InStock
76789-886 76789-888
1,1,1,5,5,5-Hexamethyl-3-phenyl-3-(trimethylsilyloxy)trisiloxane 98%
1,1,1,5,5,5-Hexamethyl-3-phenyl-3-(trimethylsilyloxy)trisiloxane
Precaución: for Research Use Only


Formula: C₁₅H₃₂O₃Si₄
MW: 372.76 g/mol
MDL Number: MFCD00053665
CAS Number: 2116-84-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty