Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiazole 98%

Supplier: AMBEED, INC
Danger

A510405-10G A510405-1G A510405-25G A510405-5G
76790-010EA 21 USD InStock
76790-010 76790-012 76790-014 76790-016
Thiazole 98%
Thiazole
Precaución: for Research Use Only


Formula: C₃H₃NS
MW: 85.13 g/mol
Boiling Pt: 116.8
Melting Pt: -34
MDL Number: MFCD00005315
CAS Number: 288-47-1
UN: 2924
ADR: 3,8 Ⅲ
Merck Index: 12,09442
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty