Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)phenyl isothiocyanate 97%

Supplier: AMBEED, INC
Danger

A521551-250MG A521551-5G A521551-1G
76790-084EA 7 USD InStock
76790-084 76790-086 77669-306
3-(Benzyloxy)phenyl isothiocyanate 97%
3-(Benzyloxy)phenyl isothiocyanate
Precaución: for Research Use Only


Formula: C₁₄H₁₁NOS
MW: 241.31 g/mol
MDL Number: MFCD00060374
CAS Number: 206559-36-6
UN: 2923
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty