Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Fluoreneacetic acid 98%

Supplier: AMBEED, INC
Warning

A522440-10G A522440-1G A522440-250MG A522440-5G
76790-088EA 21.22 USD InStock
76790-088 76790-090 76790-092 76790-094 76790-096
9-Fluoreneacetic acid 98%
9-Fluoreneacetic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
MDL Number: MFCD00013262
CAS Number: 6284-80-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty