Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-nitro-1-(trifluoromethyl)benzene 98%

Supplier: AMBEED, INC
Warning

A525825-10G A525825-1G A525825-5G
76790-120EA 49 USD InStock
76790-120 76790-122 76790-124
4-Bromo-2-nitro-1-(trifluoromethyl)benzene 98%
4-Bromo-2-nitro-1-(trifluoromethyl)benzene
Precaución: for Research Use Only


Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD11226284
CAS Number: 251115-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty