Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-7,8-dihydro-2-(methylsulfonyl)pyrido[4,3-d]pyrimidine-6(5H)-carboxylate 98%

Supplier: AMBEED, INC
Warning

A543310-1G A543310-250MG A543310-100MG
76790-230EA 110 USD InStock
76790-230 76790-232 77669-396
tert-Butyl-7,8-dihydro-2-(methylsulfonyl)pyrido[4,3-d]pyrimidine-6(5H)-carboxylate 98%
tert-Butyl-7,8-dihydro-2-(methylsulfonyl)pyrido[4,3-d]pyrimidine-6(5H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₉N₃O₄S
MDL Number: MFCD09839783
CAS Number: 259809-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty