Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutoxysilane 95%

Supplier: AMBEED, INC
Danger

A568992-100G A568992-25G A568992-500G A568992-5G
76790-332EA 77 USD InStock
76790-332 76790-334 76790-336 76790-338
Tetrabutoxysilane 95%
Tetrabutoxysilane
Precaución: for Research Use Only


Formula: C₁₆H₃₆O₄Si
MDL Number: MFCD00009432
CAS Number: 4766-57-8
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty