Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-3-Hexenoic acid 95%

Supplier: AMBEED, INC
Danger

A589132-10G A589132-25G A589132-5G
76790-464EA 20 USD InStock
76790-464 76790-466 76790-468
trans-3-Hexenoic acid 95%
trans-3-Hexenoic acid
Precaución: for Research Use Only


Formula: C₆H₁₀O₂
MDL Number: MFCD00002786
CAS Number: 1577-18-0
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty