Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylolpropane trimethacrylate 90% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A603395-100G A603395-25G A603395-500G
76790-574EA 11 USD InStock
76790-574 76790-576 76790-578
Trimethylolpropane trimethacrylate 90% stabilised
Trimethylolpropane trimethacrylate
Precaución: for Research Use Only


Formula: C₁₈H₂₆O₆
MW: 338.40 g/mol
MDL Number: MFCD00008588
CAS Number: 3290-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty