Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetracarbonyldi-µ-chlorodirhodium(I) 98%

Supplier: AMBEED, INC
Warning

A613573-100MG A613573-250MG
76790-630EA 146 USD InStock
76790-630 76790-632
Tetracarbonyldi-µ-chlorodirhodium(I) 98%
Tetracarbonyldi-µ-chlorodirhodium(I)
Precaución: for Research Use Only


Formula: C₄Cl₂O₄Rh₂
MDL Number: MFCD00135610
CAS Number: 14523-22-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty