Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethynyltoluene 95%

Supplier: AMBEED, INC
Danger

A621728-1G A621728-250MG A621728-5G
76790-664EA 13 USD InStock
76790-664 76790-666 76790-668
2-Ethynyltoluene 95%
2-Ethynyltoluene
Precaución: for Research Use Only


Formula: C₉H₈
MDL Number: MFCD03427202
CAS Number: 766-47-2
UN: 3295
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty