Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

RG-13022 98%

Supplier: AMBEED, INC
Warning

A646643-10MG A646643-1MG A646643-25MG A646643-50MG A646643-5MG
76790-834EA 20 USD InStock
76790-834 76790-836 76790-838 76790-840 76790-842
RG-13022 98%
RG-13022
Precaución: for Research Use Only


Formula: C₁₆H₁₄N₂O₂
MDL Number: MFCD00236444
CAS Number: 136831-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty