Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A664903-250MG A664903-5G A664903-1G
76790-904EA 16 USD InStock
76790-904 76790-906 77669-810
trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid 95%
trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO₄
MDL Number: MFCD01550737
CAS Number: 34771-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty